2,2,2-Trifluoroethanol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Trifluoro Ethanol, TFE
001273-250G
001273-1KG
001273-2.5KG
FLUO001273-250GEA
0
GBP
FLUO001273-250G
FLUO001273-1KG
FLUO001273-2.5KG
2,2,2-Trifluoroethanol
2,2,2-Trifluoroethanol
Formula:
C₂H₃F₃O Boiling Pt: 74.1 °C (1013 hPa) Melting Pt: –43.5 °C Density: 1,39 g/cm³ (20 °C) Flash Pt: 33 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004672 CAS Number: 75-89-8 EINECS: 200-913-6 UN: 2929 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...