1-Iodoperfluorooctane ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Perfluorooctyl iodide, Heptadecafluoro-1-iodooctane, Perfluoro-1-iodooctane

B20207.09 B20207.18
B20207.09EA 28 GBP
B20207.09 B20207.18
1-Iodoperfluorooctane ≥98%
1-Iodoperfluorooctane

Purity: 98% CAS No.: 507-63-1 Molecular Formula: C8F17I Formula Weight: 545.97 Refractive Index: 1.3290 Melting Point: 25° Boiling Point: 160-161° Beilstein Reference: 1717141 MDL No.: MFCD00001064

Formula: C₈F₁₇I
MW: 545.97 g/mol
Boiling Pt: 160…161 °C
Melting Pt: 25
Density: 2.040
Storage Temperature: Ambient
MDL Number: MFCD00001064
CAS Number: 507-63-1
EINECS: 208-079-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR