1-Iodoperfluoroheptane ≥97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Perfluoroheptyl iodide

L10206.06 L10206.14
L10206.06EA 0 GBP
L10206.06 L10206.14
1-Iodoperfluoroheptane ≥97%
1-Iodoperfluoroheptane
Formula: C₇F₁₅I
MW: 495.96 g/mol
MDL Number: MFCD00013712
CAS Number: 335-58-0
EINECS: 206-393-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR