1,4-Diiodoperfluorobutane

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Octafluoro-1,4-diiodobutane, 1,4-Diiodooctafluorobutane

004900-5G 004900-25G
FLUO004900-5GEA 0 GBP
FLUO004900-5G FLUO004900-25G
1,4-Diiodoperfluorobutane
1,4-Diiodoperfluorobutane
Formula: C₄F₈I₂
MW: 453.84 g/mol
Boiling Pt: 150 °C (1013 hPa)
Melting Pt: –3 °C
Density: 2.5466 g/cm³ (0 °C)
MDL Number: MFCD00042263
CAS Number: 375-50-8
EINECS: 206-788-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR