Trifluoroacetic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
008708-500ML
008708-50ML
008708-2.5L
FLUO008708-500MLEA
0
GBP
FLUO008708-500ML
FLUO008708-50ML
FLUO008708-2.5L
Trifluoroacetic acid
Trifluoroacetic acid
Formula:
C₂HF₃O₂ Boiling Pt: 71.8 °C (1013 hPa) Melting Pt: –15 °C Density: 1.490 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004169 CAS Number: 76-05-1 EINECS: 200-929-3 UN: 2699 ADR: 8,I Merck Index: 13,09754 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...