Trifluoroacetic acid

Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: TFA

FLUO001271-250GEA 0 GBP
FLUO001271-250G FLUO001271-5KG FLUO001271-2.5KG FLUO001271-1KG
Trifluoroacetic acid
Trifluoroacetic acid
Formula: C₂HF₃O₂
MW: 114.02 g/mol
Boiling Pt: 71.8 °C (1013 hPa)
Melting Pt: –15 °C
Density: 1.490 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 12,09812
https://uk.vwr.com/stibo/web/std.lang.all/83/92/728392.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR