1,1,1-Trifluoroacetylacetone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
001031-100G
001031-25G
FLUO001031-100GEA
0
GBP
FLUO001031-100G
FLUO001031-25G
1,1,1-Trifluoroacetylacetone
1,1,1-Trifluoroacetylacetone
Formula:
C₅H₅F₃O₂ Boiling Pt: 107 °C (1013 hPa) Density: 1,28 g/cm³ (20 °C) Flash Pt: 28 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000427 CAS Number: 367-57-7 EINECS: 206-698-5 UN: 1224 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...