Perfluorooctane
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Octadecafluorooctane
008706-100G
008706-500G
FLUO008706-100GEA
0
GBP
FLUO008706-100G
FLUO008706-500G
Perfluorooctane
Perfluorooctane
Formula:
C₈F₁₈ Boiling Pt: 102 °C (1013 hPa) Melting Pt: –25 °C Density: 1,73 g/cm³ (20 °C) |
MDL Number:
MFCD00042083 CAS Number: 307-34-6 EINECS: 206-199-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...