6-Chloro-1-hexanol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001744-50G
001744-250G
FLUO001744-50GEA
0
GBP
FLUO001744-50G
FLUO001744-250G
6-Chloro-1-hexanol
6-Chloro-1-hexanol
Formula:
C₆H₁₃ClO Boiling Pt: 110 °C (14 mmHg) Density: 1,0241 g/cm³ (20 °C) Flash Pt: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002984 CAS Number: 2009-83-8 EINECS: 217-925-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...