Trichloroacetic acid (TCA)

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

8789.1 8789.2
ROTH8789.1EA 0 GBP
ROTH8789.1 ROTH8789.2
Trichloroacetic acid (TCA)
Trichloroacetic acid (TCA)
Formula: Cl₃CCOOH
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: 1.63 g/cm³ (20 °C)
Flash Pt: 113 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
Merck Index: 13,09700
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR