1-Chlorododecane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Lauryl chloride, Dodecylchloride
24500-1L
24500-250ML
SIAL24500-1LEA
142
GBP
SIAL24500-1L
SIAL24500-250ML
1-Chlorododecane, Sigma-Aldrich®
1-Chlorododecane
Formula:
C₁₂H₂₅Cl Boiling Pt: 260 °C (1013 hPa) Melting Pt: –9 °C Density: 0.867 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013688 CAS Number: 112-52-7 EINECS: 203-981-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...