Citric acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
27487-1KG-F
27487-250G-F
27487-50G-F
SIAL27487-1KG-FEA
149
GBP
SIAL27487-1KG-F
SIAL27487-250G-F
SIAL27487-50G-F
Citric acid, Sigma-Aldrich®
Citric acid
Formula:
HOC(COOH)(CH₂COOH)₂ Boiling Pt: 310 °C (1013 hPa) Melting Pt: 149…151 °C Density: 1.665 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011669 CAS Number: 77-92-9 EINECS: 201-069-1 REACH: 01-2119457026-42-0000 Merck Index: 13,02350 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...