Triethylammonium dihydrogentrifluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Triethylamine trihydrofluoride, Triethylamine trishydrofluoride

003029-100G 003029-500G
FLUO003029-100GEA 0 GBP
FLUO003029-100G FLUO003029-500G
Triethylammonium dihydrogentrifluoride
Triethylammonium dihydrogentrifluoride
Formula: C₆H₁₈F₃N
MW: 161.21 g/mol
Density: 1.094 g/cm³ (20 °C)
MDL Number: MFCD00043294
CAS Number: 73602-61-6
EINECS: 277-550-5
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR