Triethylammonium chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Triethylamine hydrochloride

90350-1KG 90350-250G 90350-50G
SIAL90350-1KGEA 153 GBP
SIAL90350-1KG SIAL90350-250G SIAL90350-50G
Triethylammonium chloride, Sigma-Aldrich®
Triethylammonium chloride
Formula: C₆H₁₆ClN
MW: 137.65 g/mol
Density: 0.7637 g/cm³ (20 °C)
MDL Number: MFCD00012500
CAS Number: 554-68-7
EINECS: 209-067-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR