Tetrabutylammonium chloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OS-7710-25G OS-7710-100G
COBBOS-7710-25GEA 0 GBP
COBBOS-7710-25G COBBOS-7710-100G
Tetrabutylammonium chloride
Tetrabutylammonium chloride
Formula: C₁₆H₃₆ClN
MW: 277.92 g/mol
Melting Pt: 83…86 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 110
MDL Number: MFCD00011635
CAS Number: 1112-67-0
EINECS: 214-195-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR