Phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Phenylmetaboric acid, Dihydroxy(phenyl)borane, Phenyl boric acid, Benzeneboronic acid

BB-2375-25G BB-2375-100G
COBBBB-2375-25GEA 0 GBP
COBBBB-2375-25G COBBBB-2375-100G
Phenylboronic acid
Phenylboronic acid
Formula: C₆H₇BO₂
MW: 121.93 g/mol
Melting Pt: 217…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00002103
CAS Number: 98-80-6
EINECS: 202-701-9
Merck Index: 13,01068

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR