Pentafluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2,3,4,5,6-Pentafluorobenzeneboronic acid, Dihydroxy(pentafluorophenyl)borane, (Pentafluorophenyl)metaboric acid, Pentafluorobenzeneboronic acid

BB-8007-5G BB-8007-1G
COBBBB-8007-5GEA 0 GBP
COBBBB-8007-5G COBBBB-8007-1G
Pentafluorophenylboronic acid
Pentafluorophenylboronic acid
Formula: C₆H₂BF₅O₂
MW: 211.88 g/mol
MDL Number: MFCD01074663
CAS Number: 1582-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR