Palladium(II) chloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OT-0560-1G OT-0560-5G OT-0560-25G
COBBOT-0560-1GEA 0 GBP
COBBOT-0560-1G COBBOT-0560-5G COBBOT-0560-25G
Palladium(II) chloride
Palladium (II) chloride
Formula: PdCl₂
Melting Pt: 678 °C
Density: 4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003558
CAS Number: 7647-10-1
EINECS: 231-596-2
UN: 3260
ADR: 8,III
Merck Index: 13,07058

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR