Orthoboric acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
B7901-1KG
B7901-500G
SIALB7901-1KGEA
215
GBP
SIALB7901-1KG
SIALB7901-500G
Orthoboric acid, Sigma-Aldrich®
ortho-Boric acid
Formula:
B(OH)₃ Boiling Pt: 185 °C (1013 hPa) Melting Pt: 160 °C Density: 1.44 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011337 CAS Number: 10043-35-3 EINECS: 233-139-2 REACH: 01-2119486683-25 Merck Index: 13,01326 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...