Ethyl nitroacetate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Nitroethansäureethylester, Nitroacetic ethyl ester

002925-5G 002925-25G
FLUO002925-5GEA 0 GBP
FLUO002925-5G FLUO002925-25G
Ethyl nitroacetate
Ethyl nitroacetate
Formula: C₄H₇NO₄
Boiling Pt: 93…95 °C (13.3 hPa)
Density: 1,226 g/cm³ (0 °C)
MDL Number: MFCD00007403
CAS Number: 626-35-7
EINECS: 210-944-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR