Ethyl nitroacetate 98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
185170050
185170250
185171000
ACRO185170050EA
38.7
GBP
ACRO185170050
ACRO185170250
ACRO185171000
Ethyl nitroacetate 98%
Ethyl nitroacetate
Formula:
C₄H₇NO₄ Boiling Pt: 105…107 °C (25 mmHg) Density: 1.199 g/cm³ Flash Pt: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007403 CAS Number: 626-35-7 EINECS: 210-944-7 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4235 to 1.4255 (20°C, 589 nm) |
GC | ≥97.5 % |
Identification | Passes test for Chlorides |
Residue after ignition | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...