Methoxyacetyl chloride 97% stabilised

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

125700100 125700500 125702500
ACRO125700100EA 39 GBP
ACRO125700100 ACRO125700500 ACRO125702500
Methoxyacetyl chloride 97% stabilised
Methoxyacetyl chloride
Formula: C₃H₅ClO₂
Boiling Pt: 112…113 °C (760 mmHg)
Density: 1.187 g/cm³
Flash Pt: 28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000728
CAS Number: 38870-89-2
EINECS: 254-169-2
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4240 (20°C, 589 nm)
Stabilizer 0.3 % MgO
Titration RCOCl ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR