Isoxazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7607-5G
OS-7607-100G
OS-7607-25G
COBBOS-7607-5GEA
0
GBP
COBBOS-7607-5G
COBBOS-7607-100G
COBBOS-7607-25G
Isoxazole
Isoxazole
Formula:
C₃H₃NO Boiling Pt: 95 °C (1013 hPa) Density: 1,08 g/cm³ (20 °C) Flash Pt: 10 °C |
MDL Number:
MFCD00003149 CAS Number: 288-14-2 EINECS: 206-018-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...