Ethyl heptafluorobutyrate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001077-100G
001077-25G
FLUO001077-100GEA
0
GBP
FLUO001077-100G
FLUO001077-25G
Ethyl heptafluorobutyrate
Ethyl heptafluorobutyrate
Formula:
C₆H₅F₇O₂ Boiling Pt: 95 °C (1013 hPa) Density: 1.394 g/cm³ (20 °C) |
MDL Number:
MFCD00000434 CAS Number: 356-27-4 EINECS: 206-602-1 UN: 3272 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...