Ethoxyacetic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Ethoxyacetic acid

M-Clarity™ quality level = MQ200

137111-100G 137111-25G
SIAL137111-100GEA 283 GBP
SIAL137111-100G SIAL137111-25G
Ethoxyacetic acid, Sigma-Aldrich®
Ethoxyacetic acid
Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 206 °C (1013 hPa)
Density: 1.107 g/cm³ (11 °C)
MDL Number: MFCD00004310
CAS Number: 627-03-2
EINECS: 210-978-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR