4-Fluorophenyl acetate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003918-100G 003918-25G
FLUO003918-100GEA 0 GBP
FLUO003918-100G FLUO003918-25G
4-Fluorophenyl acetate
4-Fluorophenyl acetate
Formula: C₈H₇FO₂
Boiling Pt: 58.5…59.5 °C (1 mmHg)
Density: 1,17 g/cm³ (25 °C)
MDL Number: MFCD00041361
CAS Number: 405-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR