3-Phenylpropyl acetate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Phenyl-1-propyl acetate, 3-Phenylpropyl Acetate

M-Clarity™ quality level = MQ300

W289000-1KG-K W289000-5KG-K
SIALW289000-1KG-KEA 138 GBP
SIALW289000-1KG-K SIALW289000-5KG-K
3-Phenylpropyl acetate, Sigma-Aldrich®
3-Phenylpropyl acetate
Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 252.3 °C (1013 hPa)
Melting Pt: –60 °C
Density: 1.025 g/cm³ (16 °C)
MDL Number: MFCD00026216
CAS Number: 122-72-5
EINECS: 204-569-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR