3-Phenylpropyl acetate ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Phenyl-1-propyl acetate, 3-Phenylpropyl Acetate
B20878.09
B20878.18
B20878.09EA
16.9
GBP
B20878.09
B20878.18
3-Phenylpropyl acetate ≥98%
3-Phenylpropyl acetate
Purity: 98%
CAS No.: 122-72-5
Molecular Formula: CH3CO2(CH2)3C6H5
Formula Weight: 178.23
Flash Point: 130°(266°F)
Refractive Index: 1.496
Boiling Point: 252°
MDL No.: MFCD00026216
Formula:
C₁₁H₁₄O₂ MW: 178.23 g/mol Boiling Pt: 252 °C Melting Pt: –60 °C Density: 1.010 Flash Pt: 130 °C (266 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00026216 CAS Number: 122-72-5 EINECS: 204-569-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...