Palladium(II) acetate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Palladium diacetate, Palladium (II) acetate

OS-7404-5G OS-7404-25G OS-7404-1G OS-7404-100G
COBBOS-7404-5GEA 0 GBP
COBBOS-7404-5G COBBOS-7404-25G COBBOS-7404-1G COBBOS-7404-100G
Palladium(II) acetate
Palladium (II) acetate
Formula: C₄H₆O₄Pd
MW: 224.51 g/mol
Melting Pt: 205 °C
MDL Number: MFCD00012453
CAS Number: 3375-31-3
EINECS: 222-164-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR