Ethyl diethoxyacetate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
128244-100G
128244-25G
SIAL128244-100GEA
158
GBP
SIAL128244-100G
SIAL128244-25G
Ethyl diethoxyacetate, Sigma-Aldrich®
Ethyl diethoxyacetate
Formula:
C₈H₁₆O₄ Boiling Pt: 199 °C (1013 hPa) Density: 0.994 g/cm³ (18 °C) |
MDL Number:
MFCD00009231 CAS Number: 6065-82-3 EINECS: 227-999-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...