4-Nitrophenyl chloroformate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OT-0341-25G OT-0341-100G
COBBOT-0341-25GEA 0 GBP
COBBOT-0341-25G COBBOT-0341-100G
4-Nitrophenyl chloroformate
4-Nitrophenyl chloroformate
Formula: C₇H₄ClNO₄
Boiling Pt: 159 °C (19 mmHg)
Melting Pt: 78…81 °C
MDL Number: MFCD00007321
CAS Number: 7693-46-1
EINECS: 231-706-9
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR