Benzofuran

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,3-Benzofuran, Benzo[b]furan, Benzo(b)furan, 1-Benzofuran, Coumarone

OS-7561-100G OS-7561-5G OS-7561-25G OS-7561-500G
COBBOS-7561-100GEA 0 GBP
COBBOS-7561-100G COBBOS-7561-5G COBBOS-7561-25G COBBOS-7561-500G
Benzofuran
Benzofuran
Formula: C₈H₆O
MW: 118.14 g/mol
Boiling Pt: 173 °C (1013 hPa)
Melting Pt: < –18 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 50 °C
MDL Number: MFCD00005847
CAS Number: 271-89-6
EINECS: 205-982-6
Merck Index: 13,01089

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR