Piperonylic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3,4-(Methylenedioxy)benzoic acid, Benzo[d][1,3]dioxole-5-carboxylic acid, 1,3-Benzodioxole-5-carboxylic acid
OS-7549-5G
OS-7549-25G
OS-7549-100G
COBBOS-7549-5GEA
0
GBP
COBBOS-7549-5G
COBBOS-7549-25G
COBBOS-7549-100G
Piperonylic acid
Piperonylic acid
Formula:
C₈H₆O₄ MW: 166.13 g/mol Boiling Pt: 180 °C (1 mmHg) Melting Pt: 232…235 °C |
MDL Number:
MFCD00005830 CAS Number: 94-53-1 EINECS: 202-342-8 Merck Index: 12,07630 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...