8-Chloroquinoline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2105-5G
HC-2105-25G
HC-2105-1G
COBBHC-2105-5GEA
0
GBP
COBBHC-2105-5G
COBBHC-2105-25G
COBBHC-2105-1G
8-Chloroquinoline
8-Chloroquinoline
Formula:
C₉H₆ClN Boiling Pt: 288 °C (1013 hPa) Melting Pt: –20 °C Density: 1,2834 g/cm³ (14 °C) |
MDL Number:
MFCD00047618 CAS Number: 611-33-6 EINECS: 210-265-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...