7-Fluoroindole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
IN-0048-1G
IN-0048-5G
COBBIN-0048-1GEA
0
GBP
COBBIN-0048-1G
COBBIN-0048-5G
7-Fluoroindole
7-Fluoroindole
Formula:
C₈H₆FN MW: 135.14 g/mol |
MDL Number:
MFCD01074502 CAS Number: 387-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...