7-Azaindole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1H-Pyrrolo[2,3-b]pyridine
IN-0100-5G
IN-0100-25G
IN-0100-100G
COBBIN-0100-5GEA
0
GBP
COBBIN-0100-5G
COBBIN-0100-25G
COBBIN-0100-100G
7-Azaindole
7-Azaindole
Formula:
C₇H₆N₂ MW: 118.14 g/mol Boiling Pt: 384 Melting Pt: 103…106 °C Density: 1.301 g/cm³ (20 °C) |
MDL Number:
MFCD00005606 CAS Number: 271-63-6 EINECS: 205-981-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...