6-Methylisoquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6025-5G HC-6025-1G
COBBHC-6025-5GEA 0 GBP
COBBHC-6025-5G COBBHC-6025-1G
6-Methylisoquinoline
6-Methylisoquinoline
Formula: C₁₀H₉N
MDL Number: MFCD09258767
CAS Number: 42398-73-2
EINECS: 255-794-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR