6-Methylnicotinic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Methyl-3-pyridinecarboxylic acid, 6-Methylpyridine-3-carboxylic acid

OS-7595-25G OS-7595-5G
COBBOS-7595-25GEA 0 GBP
COBBOS-7595-25G COBBOS-7595-5G
6-Methylnicotinic acid
6-Methylnicotinic acid
Formula: C₇H₇NO₂
Melting Pt: 212 °C
MDL Number: MFCD00006341
CAS Number: 3222-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR