6-Methoxyindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 6-Methoxy-1H-indole, 1H-Indol-6-yl methyl ether, Indol-6-yl methyl ether

IN-0054-5G IN-0054-25G IN-0054-1G
COBBIN-0054-5GEA 0 GBP
COBBIN-0054-5G COBBIN-0054-25G COBBIN-0054-1G
6-Methoxyindole
6-Methoxyindole
Formula: C₉H₉NO
MW: 147.18 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00022780
CAS Number: 3189-13-7
EINECS: 221-689-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR