6-Hydroxynicotinic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Oxo-1,6-dihydropyridine-3-carboxylic acid, 6-Hydroxy-3-pyridinecarboxylic acid, 6-Hydroxyniacin, 6-Hydroxypyridine-3-carboxylic acid

CA-4054-25G CA-4054-100G
COBBCA-4054-25GEA 0 GBP
COBBCA-4054-25G COBBCA-4054-100G
6-Hydroxynicotinic acid
6-Hydroxynicotinic acid
Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 304…306 °C
MDL Number: MFCD00006277
CAS Number: 5006-66-6
EINECS: 225-682-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR