6-Chloronicotinamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
6-Chloropyridine-3-carboxamide
PY-7070-100G
PY-7070-25G
PY-7070-5G
COBBPY-7070-100GEA
0
GBP
COBBPY-7070-100G
COBBPY-7070-25G
COBBPY-7070-5G
6-Chloronicotinamide
6-Chloronicotinamide
Formula:
C₆H₅ClN₂O MW: 156.57 g/mol |
MDL Number:
MFCD00006242 CAS Number: 6271-78-9 EINECS: 228-456-8 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...