6-Bromoindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

IN-0045-1G IN-0045-25G IN-0045-100G IN-0045-5G
COBBIN-0045-1GEA 0 GBP
COBBIN-0045-1G COBBIN-0045-25G COBBIN-0045-100G COBBIN-0045-5G
6-Bromoindole
6-Bromoindole
Formula: C₈H₆BrN
Storage Temperature: Ambient
MDL Number: MFCD00238550
CAS Number: 52415-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR