6-Bromonicotinic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
6-Bromopyridine-3-carboxylic acid, 2-Bromo-5-pyridinecarboxylic acid, 6-Bromo-3-pyridinecarboxylic acid
CA-4051-5G
CA-4051-25G
COBBCA-4051-5GEA
0
GBP
COBBCA-4051-5G
COBBCA-4051-25G
6-Bromonicotinic acid
6-Bromonicotinic acid
Formula:
C₆H₄BrNO₂ |
MDL Number:
MFCD01927100 CAS Number: 6311-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...