5-Nitroindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Nitro-1H-indole

IN-0013-5G IN-0013-100G IN-0013-25G
COBBIN-0013-5GEA 0 GBP
COBBIN-0013-5G COBBIN-0013-100G COBBIN-0013-25G
5-Nitroindole
5-Nitroindole
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 140 °C
MDL Number: MFCD00005673
CAS Number: 6146-52-7
EINECS: 228-153-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR