5-Iodoindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

IN-0506-1G IN-0506-5G IN-0506-25G
COBBIN-0506-1GEA 0 GBP
COBBIN-0506-1G COBBIN-0506-5G COBBIN-0506-25G
5-Iodoindole
5-Iodoindole
Formula: C₈H₆IN
MW: 243.05 g/mol
MDL Number: MFCD00220065
CAS Number: 16066-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR