5-Fluoroindole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
5-Fluoro-1H-indole
IN-0047-25G
IN-0047-1G
IN-0047-5G
COBBIN-0047-25GEA
0
GBP
COBBIN-0047-25G
COBBIN-0047-1G
COBBIN-0047-5G
5-Fluoroindole
5-Fluoroindole
Formula:
C₈H₆FN Boiling Pt: 85 °C (0,5 mmHg) Melting Pt: 46 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005671 CAS Number: 399-52-0 EINECS: 206-917-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...