Indole-5-carbonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1H-Indole-5-carbonitrile, 5-Cyanoindole, 5-Cyano-1H-indole

IN-0006-5G IN-0006-25G
COBBIN-0006-5GEA 0 GBP
COBBIN-0006-5G COBBIN-0006-25G
Indole-5-carbonitrile
Indole-5-carbonitrile
Formula: C₉H₆N₂
Melting Pt: 106…108 °C
MDL Number: MFCD00005669
CAS Number: 15861-24-2
EINECS: 239-986-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR