5-Chloroindole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Chloro-1H-indole
IN-0042-25G
IN-0042-1G
IN-0042-100G
IN-0042-5G
COBBIN-0042-25GEA
0
GBP
COBBIN-0042-25G
COBBIN-0042-1G
COBBIN-0042-100G
COBBIN-0042-5G
5-Chloroindole
5-Chloroindole
Formula:
C₈H₆ClN MW: 151.6 g/mol Melting Pt: 71…72 °C |
MDL Number:
MFCD00005672 CAS Number: 17422-32-1 EINECS: 241-448-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...