5-Bromoindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

IN-0044-5G IN-0044-25G IN-0044-100G
COBBIN-0044-5GEA 0 GBP
COBBIN-0044-5G COBBIN-0044-25G COBBIN-0044-100G
5-Bromoindole
5-Bromoindole
Formula: C₈H₆BrN
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00005670
CAS Number: 10075-50-0
EINECS: 233-208-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR