5-Bromo-1H-indazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7893-1G PY-7893-25G PY-7893-5G
COBBPY-7893-1GEA 0 GBP
COBBPY-7893-1G COBBPY-7893-25G COBBPY-7893-5G
5-Bromo-1H-indazole
5-Bromo-1H-indazole
Formula: C₇H₅BrN₂
MW: 197.03 g/mol
MDL Number: MFCD00839493
CAS Number: 53857-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR